COM448633097 Combi-Blocks, Inc.

3-Ethyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

  • MDL number MFCD27936581
  • Molecular Weight : 272.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1818

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1818
Availability details for
Quantity
  • Availability
  • Add to Cart