COM448633102 Combi-Blocks, Inc.

2-(Oxan-2-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-3-carbaldehyde

  • MDL number MFCD27936586
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1823

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1823
Availability details for
Quantity
  • Availability
  • Add to Cart