COM448633107 Combi-Blocks, Inc.

2-{3-[(4-Fluorophenyl)carbonyl]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936591
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1828

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1828
Availability details for
Quantity
  • Availability
  • Add to Cart