COM448633110 Combi-Blocks, Inc.

2-(Dimethylamino)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

  • MDL number MFCD22493678
  • Cas Number : 1352819-15-8
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1831

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1831
Availability details for
Quantity
  • Availability
  • Add to Cart