COM448633117 Combi-Blocks, Inc.

{[2-Fluoro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}(isopropyl)amine

  • MDL number MFCD27936599
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1838
Availability details for
Quantity
  • Availability
  • Add to Cart