COM448633134 Combi-Blocks, Inc.

Ethyl({[2-fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl})amine

  • MDL number MFCD27936614
  • Molecular Weight : 279.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1855

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1855
Availability details for
Quantity
  • Availability
  • Add to Cart