COM448633135 Combi-Blocks, Inc.

2-(Pyrrolidin-1-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

  • MDL number MFCD27936615
  • Cas Number : 2213446-70-7
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1856
Availability details for
Quantity
  • Availability
  • Add to Cart