COM448633140 Combi-Blocks, Inc.

2-(1-Methylethoxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzaldehyde

  • MDL number MFCD22493701
  • Cas Number : 1356639-75-2
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1862
Availability details for
Quantity
  • Availability
  • Add to Cart