COM448633142 Combi-Blocks, Inc.

[2-(Benzyloxy)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol

  • MDL number MFCD27936621
  • Cas Number : 2377609-58-8
  • Molecular Weight : 340.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1864
Availability details for
Quantity
  • Availability
  • Add to Cart