COM448633147 Combi-Blocks, Inc.

tert-Butyl 4-{[2-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine-1-carboxylate

  • MDL number MFCD27936626
  • Molecular Weight : 420.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1869

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1869
Availability details for
Quantity
  • Availability
  • Add to Cart