COM448633148 Combi-Blocks, Inc.

2-{4-[(4-Methoxyphenyl)carbonyl]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936627
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1870

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1870
Availability details for
Quantity
  • Availability
  • Add to Cart