COM448633156 Combi-Blocks, Inc.

(3-Fluorophenyl)[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamine

  • MDL number MFCD27936635
  • Molecular Weight : 327.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1878

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1878
Availability details for
Quantity
  • Availability
  • Add to Cart