COM448633158 Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-indazol-3-amine

  • MDL number MFCD22571747
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1880
Availability details for
Quantity
  • Availability
  • Add to Cart