COM448633164 Combi-Blocks, Inc.

5-Methyl-2-(oxan-2-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-3-carbaldehyde

  • MDL number MFCD27936641
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1886

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1886
Availability details for
Quantity
  • Availability
  • Add to Cart