COM448633165 Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-{3-[(4-methylphenyl)carbonyl]phenyl}-1,3,2-dioxaborolane

  • MDL number MFCD27936642
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1887

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1887
Availability details for
Quantity
  • Availability
  • Add to Cart