COM448633177 Combi-Blocks, Inc.

tert-Butyl 4-[2-formyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine-1-carboxylate

  • MDL number MFCD27936652
  • Molecular Weight : 416.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1899
Availability details for
Quantity
  • Availability
  • Add to Cart