COM448633180 Combi-Blocks, Inc.

[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethoxy)phenyl]methanamine

  • MDL number MFCD27936655
  • Molecular Weight : 317.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1902
Availability details for
Quantity
  • Availability
  • Add to Cart