COM448633182 Combi-Blocks, Inc.

Ethyl 2-[2-fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-2-oxoacetate

  • MDL number MFCD27936657
  • Molecular Weight : 322.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1904

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1904
Availability details for
Quantity
  • Availability
  • Add to Cart