COM448633185 Combi-Blocks, Inc.

(3E)-4-[4-Ethoxy-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]but-3-en-2-one

  • MDL number MFCD27936660
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1907

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1907
Availability details for
Quantity
  • Availability
  • Add to Cart