COM448633190 Combi-Blocks, Inc.

(2E)-3-[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethoxy)phenyl]prop-2-enoic acid

  • MDL number MFCD27936665
  • Molecular Weight : 358.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1912

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1912
Availability details for
Quantity
  • Availability
  • Add to Cart