COM448633191 Combi-Blocks, Inc.

1-(3-Methylphenyl)-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)propan-1-one

  • MDL number MFCD27936667
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1914

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1914
Availability details for
Quantity
  • Availability
  • Add to Cart