COM448633193 Combi-Blocks, Inc.

(3E)-4-[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-6-(trifluoromethoxy)phenyl]but-3-en-2-one

  • MDL number MFCD27936669
  • Molecular Weight : 356.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1916

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1916
Availability details for
Quantity
  • Availability
  • Add to Cart