COM448633201 Combi-Blocks, Inc.

4-[3-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,2,3-thiadiazol-5-amine

  • MDL number MFCD27936676
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1924

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1924
Availability details for
Quantity
  • Availability
  • Add to Cart