COM448633205 Combi-Blocks, Inc.

tert-butyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate

  • MDL number MFCD22577263
  • Cas Number : 1446443-06-6
  • Molecular Weight : 344.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1929

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1929
Availability details for
Quantity
  • Availability
  • Add to Cart