COM448633207 Combi-Blocks, Inc.

9H-Fluoren-9-ylmethyl N-[2-(tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbamate

  • MDL number MFCD27936683
  • Cas Number : 2306405-25-2
  • Molecular Weight : 393.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1932
Availability details for
Quantity
  • Availability
  • Add to Cart