COM448633212 Combi-Blocks, Inc.

2-Methoxy-3-aminopyridine-2-boronic acid pinacol ester

  • MDL number MFCD27936688
  • Molecular Weight : 250.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1938
Availability details for
Quantity
  • Availability
  • Add to Cart