COM448633220 Combi-Blocks, Inc.

3-{[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazol-2-yl]oxy}phenol

  • MDL number MFCD27936693
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1946

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1946
Availability details for
Quantity
  • Availability
  • Add to Cart