COM448633221 Combi-Blocks, Inc.

7-Chloro-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)quinazolin-4-ol

  • MDL number MFCD27936694
  • Molecular Weight : 306.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1947

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1947
Availability details for
Quantity
  • Availability
  • Add to Cart