COM448633226 Combi-Blocks, Inc.

N-Methoxy-N-methyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxamide

  • MDL number MFCD27936699
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1952

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1952
Availability details for
Quantity
  • Availability
  • Add to Cart