COM448633227 Combi-Blocks, Inc.

tert-Butyl N-{2-[(tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]phenyl}carbamate

  • MDL number MFCD27936700
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1953

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1953
Availability details for
Quantity
  • Availability
  • Add to Cart