COM448633234 Combi-Blocks, Inc.

[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]hydrazine hydrochloride

  • MDL number MFCD27936707
  • Molecular Weight : 270.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1960

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1960
Availability details for
Quantity
  • Availability
  • Add to Cart