COM448633239 Combi-Blocks, Inc.

3-Ethoxy-4-hydroxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

  • MDL number MFCD27936712
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1966
Availability details for
Quantity
  • Availability
  • Add to Cart