COM448633240 Combi-Blocks, Inc.

{4-[(Tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]phenyl}acetic acid

  • MDL number MFCD27936713
  • Cas Number : 1498324-65-4
  • Molecular Weight : 276.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1967

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1967
Availability details for
Quantity
  • Availability
  • Add to Cart