COM448633245 Combi-Blocks, Inc.

2-(2,4-Dimethyl-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936718
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1972

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1972
Availability details for
Quantity
  • Availability
  • Add to Cart