COM448633251 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD27936725
  • Cas Number : 2259856-42-1
  • Molecular Weight : 304.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1980
Availability details for
Quantity
  • Availability
  • Add to Cart