COM448633258 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,4-dihydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD27936731
  • Molecular Weight : 239.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1988
Availability details for
Quantity
  • Availability
  • Add to Cart