COM448633263 Combi-Blocks, Inc.

Benzoic acid, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)-

  • MDL number MFCD23380411
  • Cas Number : 1383968-53-3
  • Molecular Weight : 316.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1993

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1993
Availability details for
Quantity
  • Availability
  • Add to Cart