COM448633268 Combi-Blocks, Inc.

2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenamine

  • MDL number MFCD22494396
  • Cas Number : 1126367-54-1
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1998

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1998
Availability details for
Quantity
  • Availability
  • Add to Cart