COM448633305 Combi-Blocks, Inc.

4-sec-Butylphenylboronic acid pinacol ester

  • MDL number MFCD22493526
  • Cas Number : 1268242-41-6
  • Molecular Weight : 260.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2134
Availability details for
Quantity
  • Availability
  • Add to Cart