COM448633387 Combi-Blocks, Inc.

1-{[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene]sulfonyl}piperidine

  • MDL number MFCD18915055
  • Molecular Weight : 351.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2349

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2349
Availability details for
Quantity
  • Availability
  • Add to Cart