COM448633408 Combi-Blocks, Inc.

3-Boronobenzenesulfonamide, pinacol ester

  • MDL number MFCD07781217
  • Cas Number : 486422-08-6
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2403
Availability details for
Quantity
  • Availability
  • Add to Cart