COM448633409 Combi-Blocks, Inc.

[2-Fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]hydrazine hydrochloride

  • MDL number MFCD27936745
  • Molecular Weight : 288.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2404

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2404
Availability details for
Quantity
  • Availability
  • Add to Cart