COM448633447 Combi-Blocks, Inc.

1-(4-Chlorophenyl)-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)propan-1-one

  • MDL number MFCD27936749
  • Molecular Weight : 294.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2505

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2505
Availability details for
Quantity
  • Availability
  • Add to Cart