COM448633453 Combi-Blocks, Inc.

4-[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-sulfonamido]benzoic acid

  • MDL number MFCD27936751
  • Molecular Weight : 409.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2515

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2515
Availability details for
Quantity
  • Availability
  • Add to Cart