COM448633459 Combi-Blocks, Inc.

2,3-Dichloro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD27936754
  • Cas Number : 2377608-68-7
  • Molecular Weight : 288.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2530

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2530
Availability details for
Quantity
  • Availability
  • Add to Cart