COM448633478 Combi-Blocks, Inc.

2-(Morpholin-4-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine

  • MDL number MFCD27936757
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2583

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2583
Availability details for
Quantity
  • Availability
  • Add to Cart