COM448633509 Combi-Blocks, Inc.

1-Bromo-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

  • MDL number MFCD08276318
  • Cas Number : 269410-06-2
  • Molecular Weight : 283.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2654

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2654
Availability details for
Quantity
  • Availability
  • Add to Cart