COM448633565 Combi-Blocks, Inc.

2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD27936761
  • Molecular Weight : 323.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2887

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2887
Availability details for
Quantity
  • Availability
  • Add to Cart