COM448633570 Combi-Blocks, Inc.

2-(Ethoxycarbonyl)pyrimidine-5-boronic acid pinacol ester

  • MDL number MFCD27936763
  • Cas Number : 2223043-99-8
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2892
Availability details for
Quantity
  • Availability
  • Add to Cart