COM448633571 Combi-Blocks, Inc.

1-[5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-sulfonyl]piperidine-3-carboxylic acid

  • MDL number MFCD27936764
  • Molecular Weight : 401.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2893

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2893
Availability details for
Quantity
  • Availability
  • Add to Cart